Anti-REV3L Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Lieferant: Bioss
BS-19840R-A680
BOSSBS-19840R-A680EA 890 EUR
BOSSBS-19840R-A680
Anti-REV3L Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Antikörper
Interacts with MAD2L2 to form the error prone DNA polymerase seta involved in translesion DNA synthesis.
Type: Primary
Antigen: REV3L
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 2701-2800/3130
Host: Rabbit
Isotype: IgG
Reactivity: Mouse