Anti-EGF Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-2008R-A680
BOSSBS-2008R-A680EA 899 EUR
BOSSBS-2008R-A680
Anti-EGF Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
EGF stimulates the growth of various epidermal and epithelial tissues in vivo and in vitro and of some fibroblasts in cell culture. Magnesiotropic hormone that stimulates magnesium reabsorption in the renal distal convoluted tubule via engagement of EGFR and activation of the magnesium channel TRPM6. Can induce neurite outgrowth in motoneurons of the pond snail Lymnaea stagnalis in vitro (PubMed:10964941).

Type: Primary
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR