Anti-BPNT1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)

Lieferant: Bioss
BS-20136R-A750
BOSSBS-20136R-A750EA 890 EUR
BOSSBS-20136R-A750
Anti-BPNT1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Antikörper
Converts adenosine 3'-phosphate 5'-phosphosulfate (PAPS) to adenosine 5'-phosphosulfate (APS) and 3'(2')-phosphoadenosine 5'- phosphate (PAP) to AMP. Has 1000-fold lower activity towards inositol 1,4-bisphosphate (Ins(1,4)P2) and inositol 1,3,4-trisphosphate (Ins(1,3,4)P3), but does not hydrolyse Ins(1)P, Ins(3,4)P2, Ins(1,3,4,5)P4 or InsP6.

Type: Primary
Antigen: BPNT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 2-100/308
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR