Anti-DH-5A Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-2033R
BOSSBS-2033REA 704 EUR
BOSSBS-2033R
Anti-DH-5A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DH-5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR