Anti-DH-5A Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-2033R-A488
BOSSBS-2033R-A488EA 876 EUR
BOSSBS-2033R-A488
Anti-DH-5A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
This antibody eacts with Escherichia coli HD5a (E.coli HD5a), Escherichia coli is a gram negative bacillus that belongs to a larger group of Enterobacteriae - bacteria that inhabit the gastrointestinal tract. Although usually a harmless resident of the gut, some strains have the potential to cause serious problems, especially where there is an immature immune system or immunosuppression, or where the subtype of organism has acquired the ability to produce pathogenic toxins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DH-5A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR