Anti-DH-5A Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-2033R-CY5.5
BOSSBS-2033R-CY5.5EA 883 EUR
BOSSBS-2033R-CY5.5
Anti-DH-5A Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
This antibody eacts with Escherichia coli HD5a (E.coli HD5a), Escherichia coli is a gram negative bacillus that belongs to a larger group of Enterobacteriae - bacteria that inhabit the gastrointestinal tract. Although usually a harmless resident of the gut, some strains have the potential to cause serious problems, especially where there is an immature immune system or immunosuppression, or where the subtype of organism has acquired the ability to produce pathogenic toxins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DH-5A
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria