Anti-AQP9 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-2060R-CY5.5
BOSSBS-2060R-CY5.5EA 909 EUR
BOSSBS-2060R-CY5.5
Anti-AQP9 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Forms a channel with a broad specificity. Mediates passage of a wide variety of non-charged solutes including carbamides, polyols, purines, and pyrimidines in a phloretin- and mercury-sensitive manner, whereas amino acids, cyclic sugars, Na(+), K(+), Cl(-), and deprotonated monocarboxylates are excluded. Also permeable to urea and glycerol.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: AQP9
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat