Anti-Melatonin Receptor 1A Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-20768R
BOSSBS-20768REA 703 EUR
BOSSBS-20768R
Anti-Melatonin Receptor 1A Rabbit Polyclonal Antibody
Antikörper
High affinity receptor for melatonin. Likely to mediates the reproductive and circadian actions of melatonin. The activity of this receptor is mediated by pertussis toxin sensitive G proteins that inhibit adenylate cyclase activity.

Type: Primary
Antigen: Melatonin Receptor 1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Public Immunogen Range: 121-200/350
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR