Anti-Integrin alpha 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-2095R-A750
BOSSBS-2095R-A750EA 899 EUR
BOSSBS-2095R-A750
Anti-Integrin alpha 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Integrin alpha-1/beta-1 is a receptor for laminin and collagen. It recognises the proline-hydroxylated sequence G-F-P-G-E-R in collagen. Involved in anchorage-dependent, negative regulation of EGF-stimulated cell growth.

Type: Primary
Antigen: Integrin alpha 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1001-1179/1179
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR