Anti-ITGA1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-2095R-CY5
BOSSBS-2095R-CY5EA 909 EUR
BOSSBS-2095R-CY5
Anti-ITGA1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Integrin alpha-1/beta-1 is a receptor for laminin and collagen. It recognizes the proline-hydroxylated sequence G-F-P-G-E-R in collagen. Involved in anchorage-dependent, negative regulation of EGF-stimulated cell growth.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ITGA1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR