Anti-SCYLV Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-2121R-A680
BOSSBS-2121R-A680EA 890 EUR
BOSSBS-2121R-A680
Anti-SCYLV Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Yellow leaf disease of sugarcane (Saccharum spp.) is an important and widely spread disease causing severe yield losses, ranging from 20 to 40 % in susceptible cultivars. It is an emerging viral disease whose causal agent is a Polerovirus, the Sugarcane yellow leaf virus (SCYLV) transmitted by aphids.

Type: Primary
Antigen: SCYLV
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Virus, Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR