Anti-DKK1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-2162R-A750
BOSSBS-2162R-A750EA 899 EUR
BOSSBS-2162R-A750
Anti-DKK1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Antagonizes canonical Wnt signaling by inhibiting LRP5/6 interaction with Wnt and by forming a ternary complex with the transmembrane protein KREMEN that promotes internalisation of LRP5/6. DKKs play an important role in vertebrate development, where they locally inhibit Wnt regulated processes such as antero-posterior axial patterning, limb development, somitogenesis and eye formation. In the adult, Dkks are implicated in bone formation and bone disease, cancer and Alzheimer disease.

Type: Primary
Antigen: DKK1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-266/266
Host: Rabbit
Isotype: IgG
Reactivity: Human, Dog
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR