Anti-NQO1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-2184R-A555
BOSSBS-2184R-A555EA 901 EUR
BOSSBS-2184R-A555
Anti-NQO1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The enzyme apparently serves as a quinone reductase in connection with conjugation reactions of hydroquinons involved in detoxification pathways as well as in biosynthetic processes such as the vitamin K-dependent gamma-carboxylation of glutamate residues in prothrombin synthesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NQO1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR