Anti-ARHGEF2 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-2201R-CY7
BOSSBS-2201R-CY7EA 929 EUR
BOSSBS-2201R-CY7
Anti-ARHGEF2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
GEF-H1 is a Rho guanine nucleotide exchange factor that localizes to microtubules and regulates Rho activity in response to microtubule destabilization (loss of interaction between GEF-H1 and microtubules leads to activation of Rho). This protein has also been shown to localize to tight junctions and modulate polarized cell permeability.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARHGEF2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR