Anti-ARHGEF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-2201R-HRP
BOSSBS-2201R-HRPEA 929 EUR
BOSSBS-2201R-HRP
Anti-ARHGEF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
GEF-H1 is a Rho guanine nucleotide exchange factor that localizes to microtubules and regulates Rho activity in response to microtubule destabilization (loss of interaction between GEF-H1 and microtubules leads to activation of Rho). This protein has also been shown to localize to tight junctions and modulate polarized cell permeability.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ARHGEF2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR