Anti-DHRS3 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-2286R-CY5.5
BOSSBS-2286R-CY5.5EA 893 EUR
BOSSBS-2286R-CY5.5
Anti-DHRS3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
DHRS3 belongs to the short-chain dehydrogenases/reductases (SDR) family. It catalyzes the reduction of all-trans-retinal to all-trans-retinol in the presence of NADPH. DHRS3 is located in a region of chromosome 1 which is often deleted in aggressive neuroblastoma tumors. There are two named isoforms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DHRS3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR