Anti-HBV D pre-S1/S2 Protein Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-2328R
BOSSBS-2328REA 1090 EUR
BOSSBS-2328R
Anti-HBV D pre-S1/S2 Protein Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HBV D pre-S1/S2 Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR