Anti-PGF2 alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-2341R-A750
BOSSBS-2341R-A750EA 890 EUR
BOSSBS-2341R-A750
Anti-PGF2 alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Luteolytic prostaglandin that induces parturition. Potent vasoconstrictor of pulmonary, coronary, and cerebral arteriole beds. A positive feedback loop between endometrial PG F2 and luteal oxytocin is responsible for completion of luteolysis.Luteolytic prostaglandin that induces parturition. Potent vasoconstrictor of pulmonary, coronary, and cerebral arteriole beds. Also stimulates contraction of smooth muscle, such as in bronchi, with possible involvement in asthma. This effect is at least partially mediated through tachykinin release.

Type: Primary
Antigen: PGF2 alpha
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Rat; Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR