Anti-GNMT Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-2396R-CY3
BOSSBS-2396R-CY3EA 900 EUR
BOSSBS-2396R-CY3
Anti-GNMT Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Catalyzes the methylation of glycine by using S-adenosylmethionine (AdoMet) to form N-methylglycine (sarcosine) with the concomitant production of S-adenosylhomocysteine (AdoHcy). Possible crucial role in the regulation of tissue concentration of AdoMet and of metabolism of methionine.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GNMT
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat