Anti-PPIL1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-2398R-A750
BOSSBS-2398R-A750EA 890 EUR
BOSSBS-2398R-A750
Anti-PPIL1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Peptidyl-prolyl cis-trans isomerase (PPIL1) is a member of the cyclophilin family of peptidylprolyl isomerases (PPIases). The cyclophilins are a highly conserved, ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV1 virions. Based on similarity to other PPIases, this protein could accelerate the folding of proteins and might catalyse the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.

Type: Primary
Antigen: PPIL1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-130/166
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR