Anti-CYP7A1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-2399R
BOSSBS-2399REA 752 EUR
BOSSBS-2399R
Anti-CYP7A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP7A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR