Anti-MPG Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-2412R-A555
BOSSBS-2412R-A555EA 893 EUR
BOSSBS-2412R-A555
Anti-MPG Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
MPG (N-methylpurine DNA glycosylase, or 3-methyladenine (3MeA) DNA glycosylase) repairs the 3MeA lethal lesion blocking DNA replication in Escherichia coli. Analysis of human cDNA libraries found a cDNA that would hybridize to human genomic DNA but not to E. coli or yeast DNA. This MPG gene has been mapped to human chromosome 16 by analysis of a panel of DNAs from mouse/human and hamster/human hybrid cell lines.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MPG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR