Anti-APNG/MPG Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-2412R-A750
BOSSBS-2412R-A750EA 890 EUR
BOSSBS-2412R-A750
Anti-APNG/MPG Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
MPG (N-methylpurine DNA glycosylase, or 3-methyladenine (3MeA) DNA glycosylase) repairs the 3MeA lethal lesion blocking DNA replication in Escherichia coli. Analysis of human cDNA libraries found a cDNA that would hybridise to human genomic DNA but not to E. coli or yeast DNA. This MPG gene has been mapped to human chromosome 16 by analysis of a panel of DNAs from mouse/human and hamster/human hybrid cell lines.

Type: Primary
Antigen: APNG/MPG
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/298
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR