Anti-NCR3 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-2418R-CY5.5
BOSSBS-2418R-CY5.5EA 893 EUR
BOSSBS-2418R-CY5.5
Anti-NCR3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Cytotoxicity-activating receptor that contributes to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis. Engagement of NCR3 by BAG6 also promotes dendritic cell (DC) maturation, both through killing those DCs that did not properly acquire a mature phenotype, and inducing NK cells to release TNFA and IFNG, which promotes DC maturation.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NCR3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR