Anti-CD158E Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Bioss
BS-2421R-HRP
BOSSBS-2421R-HRPEA 900 EUR
BOSSBS-2421R-HRP
Anti-CD158E Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
KIR3DL1 (NKB1, CD158e1) is expressed on a subset of natural killer cells and T cells. NKB1 is a 70 kD member of the immunoglobulin superfamily that is expressed at varying levels among individuals. NKB1 is a type I membrane protein containing two immunoglobulin C2 type domains. The interaction of NKB1 with specific HLA B antigens on a target cell (the HLA Bw4 allele, for example) inhibits cytotoxicity and prevents target cell lysis and death. The interactions between KIR and MHC class I are thought to be important in NK and T cell regulation following antigen stimulation. The absence of ligands for KIRs may lower the threshold for activation through activating receptors and increase inflammation and susceptibility to autoimmune disease.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: CD158e
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse, Rat