Anti-Nuclear Matrix Protein p84/THOC1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-2448R-CY5.5
BOSSBS-2448R-CY5.5EA 893 EUR
BOSSBS-2448R-CY5.5
Anti-Nuclear Matrix Protein p84/THOC1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
This protein is part of the THO complex (THOC). The THO complex is recruited to transcribed genes and travels with the RNA polymerase during elongation. It may physically link proteins that function in transcription and in RNA export and RNA processing.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Nuclear Matrix Protein p84/THOC1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR