Anti-Nuclear Matrix Protein p84/THOC1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-2448R-HRP
BOSSBS-2448R-HRPEA 893 EUR
BOSSBS-2448R-HRP
Anti-Nuclear Matrix Protein p84/THOC1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
This protein is part of the THO complex (THOC). The THO complex is recruited to transcribed genes and travels with the RNA polymerase during elongation. It may physically link proteins that function in transcription and in RNA export and RNA processing.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: Nuclear Matrix Protein p84/THOC1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR