Anti-ZNF217 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-2510R-CY7
BOSSBS-2510R-CY7EA 876 EUR
BOSSBS-2510R-CY7
Anti-ZNF217 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
ZNF217 (Zinc-finger protein 217) is a Kruppel-like zinc-finger protein which functions as a transcriptional repressor protein and forms a complex with a number of proteins including CoREST, BHC110/LSD1, HDAC2 and CtBP1. ZNF217 has been shown to repress E-cadherin promoter activity. ZNF217 is overexpressed in a number of cancers including breast and ovarian. ZNF217 and its associated proteins have been implicated in a novel pathway that is involved in cancer progression.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF217
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR