Anti-VCAN Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: Bioss
BS-2533R-FITC
BOSSBS-2533R-FITCEA 893 EUR
BOSSBS-2533R-FITC
Anti-VCAN Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
May play a role in intercellular signaling and in connecting cells with the extracellular matrix. May take part in the regulation of cell motility, growth and differentiation. Binds hyaluronic acid.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: VCAN
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat