Anti-GLUT12 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-2540R-A680
BOSSBS-2540R-A680EA 899 EUR
BOSSBS-2540R-A680
Anti-GLUT12 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
GLUT12 belongs to a family of transporters that catalyse the uptake of sugars through facilitated diffusion. Thirteen different types of glucose/fructose transport carrier proteins designated as Glut 1-13 facilitate glucose/fructose transport across the cell membrane. Individual members of the Glut family have predicted secondary structure characteristic of 12 membrane spanning domains of other transport carriers.

Type: Primary
Antigen: GLUT12
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 251-350/617
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR