Anti-SLC2A12 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-2540R-CY5.5
BOSSBS-2540R-CY5.5EA 909 EUR
BOSSBS-2540R-CY5.5
Anti-SLC2A12 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
GLUT12 belongs to a family of transporters that catalyze the uptake of sugars through facilitated diffusion. Thirteen different types of glucose/fructose transport carrier proteins designated as Glut 1-13 facilitate glucose/fructose transport across the cell membrane. Individual members of the Glut family have predicted secondary structure characteristic of 12 membrane spanning domains of other transport carriers.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC2A12
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:More Product Information
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR