Anti-SLC2A12 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-2540R-CY7
BOSSBS-2540R-CY7EA 909 EUR
BOSSBS-2540R-CY7
Anti-SLC2A12 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
GLUT12 belongs to a family of transporters that catalyze the uptake of sugars through facilitated diffusion. Thirteen different types of glucose/fructose transport carrier proteins designated as Glut 1-13 facilitate glucose/fructose transport across the cell membrane. Individual members of the Glut family have predicted secondary structure characteristic of 12 membrane spanning domains of other transport carriers.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLC2A12
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat