Anti-CYP1A2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-2589R-A350
BOSSBS-2589R-A350EA 901 EUR
BOSSBS-2589R-A350
Anti-CYP1A2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Cytochromes P450 are a group of heme-thiolate monooxygenases. In liver microsomes, this enzyme is involved in an NADPH-dependent electron transport pathway. It oxidizes a variety of structurally unrelated compounds, including steroids, fatty acids, and xenobiotics. Most active in catalyzing 2-hydroxylation. Caffeine is metabolized primarily by cytochrome CYP1A2 in the liver through an initial N3-demethylation. Also acts in the metabolism of aflatoxin B1 and acetaminophen. Participates in the bioactivation of carcinogenic aromatic and heterocyclic amines. Catalizes the N-hydroxylation ofMore Product Information