Anti-CYP1A2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-2589R-A488
BOSSBS-2589R-A488EA 901 EUR
BOSSBS-2589R-A488
Anti-CYP1A2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Cytochromes P450 are a group of heme-thiolate monooxygenases. In liver microsomes, this enzyme is involved in an NADPH-dependent electron transport pathway. It oxidizes a variety of structurally unrelated compounds, including steroids, fatty acids, and xenobiotics. Most active in catalyzing 2-hydroxylation. Caffeine is metabolized primarily by cytochrome CYP1A2 in the liver through an initial N3-demethylation. Also acts in the metabolism of aflatoxin B1 and acetaminophen. Participates in the bioactivation of carcinogenic aromatic and heterocyclic amines. Catalizes the N-hydroxylation ofMore Product Information
Bestellen

TECHNISCHE DATEN

Antigen-Symbol CYP1A2
Environmentally Preferable
Antigen-Name Cytochrome P450, Family 1, Subfamily A, Polypeptide 2
Antigen-Synonyme BMD|TU15B|RP50|P3-450|P450(PA)|CP12|VMD2|ARB|BEST
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 488
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 1544
Isotyp IgG
ImmunoChemie Yes
Immunogen 245-285/513
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR