Anti-CYP1A2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-2589R-CY3
BOSSBS-2589R-CY3EA 909 EUR
BOSSBS-2589R-CY3
Anti-CYP1A2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Cytochromes P450 are a group of heme-thiolate monooxygenases. In liver microsomes, this enzyme is involved in an NADPH-dependent electron transport pathway. It oxidizes a variety of structurally unrelated compounds, including steroids, fatty acids, and xenobiotics. Most active in catalyzing 2-hydroxylation. Caffeine is metabolized primarily by cytochrome CYP1A2 in the liver through an initial N3-demethylation. Also acts in the metabolism of aflatoxin B1 and acetaminophen. Participates in the bioactivation of carcinogenic aromatic and heterocyclic amines. Catalizes the N-hydroxylation of heterocyclic amines and the O-deethylation of phenacetin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP1A2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR