Anti-IL17RA/CD217 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-2606R-A680
BOSSBS-2606R-A680EA 899 EUR
BOSSBS-2606R-A680
Anti-IL17RA/CD217 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Receptor for IL17A, IL17F and, in dimer with IL17RE, for IL17C. Binds its IL17A ligand with low affinity, suggesting that additional components are involved in IL17A-induced signaling.
Type: Primary
Antigen: IL17RA/CD217
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/866
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse