Anti-IL17RA Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-2606R-CY7
BOSSBS-2606R-CY7EA 909 EUR
BOSSBS-2606R-CY7
Anti-IL17RA Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Receptor for IL17A, IL17F and, in dimer with IL17RE, for IL17C. Binds its IL17A ligand with low affinity, suggesting that additional components are involved in IL17A-induced signaling.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL17RA
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR