Anti-S100 A13 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-2617R-A555
BOSSBS-2617R-A555EA 893 EUR
BOSSBS-2617R-A555
Anti-S100 A13 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Plays a role in the export of proteins that lack a signal peptide and are secreted by an alternative pathway. Binds two calcium ions per subunit. Binds one copper ion. Binding of one copper ion does not interfere with calcium binding. Required for the copper-dependent stress-induced export of IL1A and FGF1. The calcium-free protein binds to lipid vesicles containing phosphatidylserine, but not to vesicles containing phosphatidylcholine (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: S100 A13
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR