Anti-CD158 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-2643R-A680
BOSSBS-2643R-A680EA 890 EUR
BOSSBS-2643R-A680
Anti-CD158 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Receptor on natural killer (NK) cells for HLA-C alleles. Inhibits the activity of NK cells thus preventing cell lysis.
Type: Primary
Antigen: CD158
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/377
Host: Rabbit
Isotype: IgG
Reactivity: Human