Anti-SPHK1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-2652R-CY3
BOSSBS-2652R-CY3EA 909 EUR
BOSSBS-2652R-CY3
Anti-SPHK1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Sphingosine-1-phosphate (SPP) is a novel lipid messenger with both intracellular and extracellular functions. Intracellularly, it regulates proliferation and survival, and extracellularly, it is a ligand for EDG1 (MIM 601974). Various stimuli increase cellular levels of SPP by activation of sphingosine kinase (SPHK), the enzyme that catalyzes the phosphorylation of sphingosine. Competitive inhibitors of SPHK block formation of SPP and selectively inhibit cellular proliferation induced by a variety of factors, including platelet-derived growth factor (e.g., MIM 173430) and serum.[supplied by OMIM].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPHK1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR