Anti-SPHK2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-2653R-A750
BOSSBS-2653R-A750EA 899 EUR
BOSSBS-2653R-A750
Anti-SPHK2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Catalyses the phosphorylation of sphingosine to form sphingosine 1-phosphate (SPP), a lipid mediator with both intra- and extracellular functions. Also acts on D-erythro-dihydrosphingosine, D-erythro-sphingosine and L-threo-dihydrosphingosine. Binds phosphoinositides.
Type: Primary
Antigen: SPHK2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/654
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat