Anti-SPHK2 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-2653R-CY3
BOSSBS-2653R-CY3EA 901 EUR
BOSSBS-2653R-CY3
Anti-SPHK2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Catalyzes the phosphorylation of sphingosine to form sphingosine 1-phosphate (SPP), a lipid mediator with both intra- and extracellular functions. Also acts on D-erythro-dihydrosphingosine, D-erythro-sphingosine and L-threo-dihydrosphingosine. Binds phosphoinositides.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SPHK2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat