Anti-LTB4R Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-2654R-A488
BOSSBS-2654R-A488EA 901 EUR
BOSSBS-2654R-A488
Anti-LTB4R Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Receptor for extracellular ATP >UTP and ADP. The activity of this receptor is mediated by G proteins which activate a phosphatidylinositol-calcium second messenger system. May be the cardiac P2Y receptor involved in the regulation of cardiac muscle contraction through modulation of L-type calcium currents. Is a receptor for leukotriene B4, a potent chemoattractant involved in inflammation and immune response.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LTB4R
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR