Anti-SIGLEC10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-2706R-A750
BOSSBS-2706R-A750EA 890 EUR
BOSSBS-2706R-A750
Anti-SIGLEC10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
SIGLEC10 is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It preferentially binds to alpha2,3- or 2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. In the immune response, may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. SIGLEC10 interacts with PTPN6/SHP-1 upon phosphorylation. The protein isMore Product Information