Anti-GSTp1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-2735R-A750
BOSSBS-2735R-A750EA 371 EUR
BOSSBS-2735R-A750
Anti-GSTp1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles. Can metabolize 1-chloro-2,4-dinitrobenzene. Regulates negatively CDK5 activity via p25/p35 translocation to prevent neurodegeneration (By similarity).

Type: Primary
Antigen: GSTp1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 121-210/210
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR