Anti-GSTP1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-2735R-CY5.5
BOSSBS-2735R-CY5.5EA 374 EUR
BOSSBS-2735R-CY5.5
Anti-GSTP1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles. Can metabolize 1-chloro-2,4-dinitrobenzene. Regulates negatively CDK5 activity via p25/p35 translocation to prevent neurodegeneration (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GSTp1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR