Anti-FANCD2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-2738R-HRP
BOSSBS-2738R-HRPEA 893 EUR
BOSSBS-2738R-HRP
Anti-FANCD2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Fanconi Anemia (FANC) is a human autosomal-recessive cancer susceptibility disorder characterized by congenital defects, progressive bone marrow failure, and cellular hypersensitivity to mitomycin C (MMC). The FANC subunit D2 protein is vital for cellular resistance to DNA cross-linking and the arrest of DNA synthesis after ionizing radiation.DNA damage activates the monoubiquitination of FANC D2, targeting nuclear foci containing the BRCA 1 protein.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: FANCD2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR