Anti-ACACA Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-2745R-A750
BOSSBS-2745R-A750EA 890 EUR
BOSSBS-2745R-A750
Anti-ACACA Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Catalyses the rate-limiting reaction in the biogenesis of long-chain fatty acids. Carries out three functions: biotin carboxyl carrier protein, biotin carboxylase and carboxyltransferase.

Type: Primary
Antigen: ACACA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 2307-2355/2458
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR