Anti-DEFA4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-2799R-A350
BOSSBS-2799R-A350EA 893 EUR
BOSSBS-2799R-A350
Anti-DEFA4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several alpha defensin genes are clustered on chromosome 8. This gene differs from other genes of this family by an extra 83-base segment that is apparently the result of a recent duplication withinMore Product Information