Anti-DEFA4 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-2799R-CY3
BOSSBS-2799R-CY3EA 900 EUR
BOSSBS-2799R-CY3
Anti-DEFA4 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several alpha defensin genes are clustered on chromosome 8. This gene differs from other genes of this family by an extra 83-base segment that is apparently the result of a recent duplication within the coding region. The protein encoded by this gene, defensin, alpha 4, is found in the neutrophils; it exhibits corticostatic activity and inhibits corticotropin stimulated corticosterone production. [provided by RefSeq].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DEFA4
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR